Assay Details
| Gene Symbol: Bruce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275532.2 | NP_001262461.1 | 7 - 8 | 3791 | 63 | |
| NM_001275533.2 | NP_001262462.1 | 7 - 8 | 3791 | 63 | ||
| NM_001275534.2 | NP_001262463.1 | 7 - 8 | 3791 | 63 | ||
| NM_141738.5 | NP_649995.2 | 7 - 8 | 3791 | 63 | ||
| GenBank mRNA | AF517634.1 | - | 7 - 8 | 3788 | 63 | |
More Information
Set Membership: |
|

Bruce
Genomic Map