Assay Details
| Gene Symbol: Rfx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104268.2 | NP_001097738.2 | 10 - 11 | 3135 | 63 | |
| NM_001260092.2 | NP_001247021.1 | 9 - 10 | 3201 | 63 | ||
| NM_001260093.2 | NP_001247022.2 | 8 - 9 | 2728 | 63 | ||
| NM_001260094.2 | NP_001247023.2 | 9 - 10 | 3225 | 63 | ||
| NM_001275535.1 | NP_001262464.1 | 7 - 8 | 2794 | 63 | ||
| NM_141742.3 | NP_649999.2 | 10 - 11 | 3159 | 63 | ||
More Information
Set Membership: |
|

Rfx
Genomic Map