Assay Details
| Gene Symbol: Rfx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260094.2 | NP_001247023.2 | 2 - 3 | 841 | 63 | |
| NM_141742.3 | NP_649999.2 | 2 - 3 | 841 | 63 | ||
More Information
Set Membership: |
|

Rfx
Genomic Map