Assay Details
| Gene Symbol: Rfx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104268.2 | NP_001097738.2 | 4 - 5 | 1289 | 88 | |
| NM_001260092.2 | NP_001247021.1 | 4 - 5 | 1289 | 88 | ||
| NM_001260093.2 | NP_001247022.2 | 2 - 3 | 882 | 88 | ||
| NM_001260094.2 | NP_001247023.2 | 4 - 5 | 1313 | 88 | ||
| NM_001275535.1 | NP_001262464.1 | 2 - 3 | 882 | 88 | ||
| NM_141742.3 | NP_649999.2 | 4 - 5 | 1313 | 88 | ||
More Information
Set Membership: |
|

Rfx
Genomic Map