Assay Details
| Gene Symbol: Cyp12e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141746.4 | NP_650003.4 | 4 - 5 | 543 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp12e1| Gene Symbol: Cyp12e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141746.4 | NP_650003.4 | 4 - 5 | 543 | 92 | |
Set Membership: |
|