Assay Details
| Gene Symbol: TTLL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141764.4 | NP_650021.1 | 2 - 3 | 1319 | 84 | |
| GenBank mRNA | AY061104.1 | - | 2 - 3 | 1316 | 84 | |
More Information
Set Membership: |
|

TTLL15
Genomic Map