Assay Details
| Gene Symbol: TTLL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141764.4 | NP_650021.1 | 3 - 4 | 1531 | 64 | |
| GenBank mRNA | AY061104.1 | - | 3 - 4 | 1528 | 64 | |
More Information
Set Membership: |
|

TTLL15
Genomic Map