Assay Details
| Gene Symbol: Cad86C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260102.1 | NP_001247031.1 | 3 - 4 | 1144 | 85 | |
| NM_001260104.1 | NP_001247033.1 | 3 - 4 | 1144 | 85 | ||
| NM_001260105.1 | NP_001247034.1 | 3 - 4 | 1800 | 85 | ||
| NM_176458.4 | NP_788635.3 | 3 - 4 | 1800 | 85 | ||
More Information
Set Membership: |
|

Cad86C
Genomic Map