Assay Details
| Gene Symbol: Tengl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141781.2 | NP_650038.2 | 2 - 3 | 480 | 137 | |
| GenBank mRNA | BT022328.1 | - | 2 - 3 | 480 | 137 | |
More Information
Set Membership: |
|

Tengl4
Genomic Map