Assay Details
| Gene Symbol: Ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300355.1 | NP_001287284.1 | 3 - 4 | 1303 | 62 | |
| NM_141811.2 | NP_650068.1 | 3 - 4 | 1371 | 62 | ||
| GenBank mRNA | AY128454.1 | - | 3 - 4 | 1371 | 62 | |
More Information
Set Membership: |
|

Ranbp9
Genomic Map