Assay Details
| Gene Symbol: Elp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275548.1 | NP_001262477.1 | 4 - 5 | 3365 | 69 | |
| NM_001275549.1 | NP_001262478.1 | 4 - 5 | 3374 | 69 | ||
| NM_141841.2 | NP_650098.1 | 4 - 5 | 3362 | 69 | ||
More Information
Set Membership: |
|

Elp1
Genomic Map