Assay Details
| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275561.1 | NP_001262490.1 | 3 - 4 | 378 | 64 | |
| NM_141877.2 | NP_650134.1 | 3 - 4 | 378 | 64 | ||
| GenBank mRNA | AY119214.1 | - | 3 - 4 | 357 | 64 | |
More Information
Set Membership: |
|

GC1
Genomic Map