Assay Details
| Gene Symbol: CG14740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141895.3 | NP_650152.1 | 2 - 3 | 345 | 85 | |
| GenBank mRNA | AY058419.1 | - | 2 - 3 | 293 | 85 | |
More Information
Set Membership: |
|

CG14740
Genomic Map