Assay Details
Gene Symbol: CG34402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104286.2 | NP_001097756.1 | 1 - 2 | 148 | 69 | |
NM_001300376.1 | NP_001287305.1 | 1 - 2 | 115 | 69 | ||
GenBank mRNA | DQ015970.1 | - | 1 - 2 | 109 | 69 |

More Information
Set Membership: |
![]() ![]() |
