Assay Details
Gene Symbol: CG34402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104286.2 | NP_001097756.1 | 5 - 6 | 1538 | 62 | |
NM_001104288.2 | NP_001097758.1 | 3 - 4 | 1388 | 62 | ||
NM_001300376.1 | NP_001287305.1 | 5 - 6 | 1505 | 62 | ||
GenBank mRNA | BT023111.1 | - | 3 - 4 | 1388 | 62 | |
DQ015970.1 | - | 5 - 6 | 1499 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
