Assay Details
| Gene Symbol: CG34402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104286.2 | NP_001097756.1 | 6 - 7 | 1741 | 75 | |
| NM_001104288.2 | NP_001097758.1 | 4 - 5 | 1591 | 75 | ||
| NM_001300376.1 | NP_001287305.1 | 6 - 7 | 1708 | 75 | ||
| GenBank mRNA | BT023111.1 | - | 4 - 5 | 1591 | 75 | |
| DQ015970.1 | - | 6 - 7 | 1702 | 75 | ||
More Information
Set Membership: |
|

CG34402
Genomic Map