Assay Details
| Gene Symbol: Sccpdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141933.2 | NP_650190.1 | 1 | 1481 | 66 | |
| GenBank mRNA | BT024191.1 | - | 1 | 1479 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sccpdh2| Gene Symbol: Sccpdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141933.2 | NP_650190.1 | 1 | 1481 | 66 | |
| GenBank mRNA | BT024191.1 | - | 1 | 1479 | 66 | |
Set Membership: |
|