Assay Details
| Gene Symbol: pps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260146.2 | NP_001247075.1 | 6 - 7 | 3766 | 62 | |
| NM_141936.2 | NP_650193.1 | 6 - 7 | 3760 | 62 | ||
More Information
Set Membership: |
|

pps
Genomic Map