Assay Details
| Gene Symbol: pps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260146.2 | NP_001247075.1 | 7 - 8 | 5712 | 57 | |
| NM_141936.2 | NP_650193.1 | 7 - 8 | 5706 | 57 | ||
More Information
Set Membership: |
|

pps
Genomic Map