Assay Details
| Gene Symbol: CG5961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202282.1 | NP_001189211.1 | 2 - 3 | 739 | 76 | |
| NM_001300378.1 | NP_001287307.1 | 2 - 3 | 929 | 76 | ||
| NM_141949.4 | NP_650206.1 | 2 - 3 | 934 | 76 | ||
| GenBank mRNA | AY069677.1 | - | 2 - 3 | 897 | 76 | |
More Information
Set Membership: |
|

CG5961
Genomic Map