Assay Details
| Gene Symbol: CG5608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141951.1 | NP_650208.1 | 4 - 5 | 1023 | 59 | |
| GenBank mRNA | BT050577.1 | - | 4 - 5 | 1023 | 59 | |
More Information
Set Membership: |
|

CG5608
Genomic Map