Assay Details
| Gene Symbol: Not10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316562.1 | NP_001303491.1 | 3 - 4 | 1242 | 80 | |
| NM_141958.4 | NP_650215.1 | 3 - 4 | 1242 | 80 | ||
More Information
Set Membership: |
|

Not10
Genomic Map