Assay Details
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141966.4 | NP_650223.1 | 3 - 4 | 643 | 63 | |
| GenBank mRNA | BT003464.1 | - | 3 - 4 | 642 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnmt| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141966.4 | NP_650223.1 | 3 - 4 | 643 | 63 | |
| GenBank mRNA | BT003464.1 | - | 3 - 4 | 642 | 63 | |
Set Membership: |
|