Assay Details
| Gene Symbol: Lkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170135.2 | NP_001163606.1 | 2 - 3 | 1447 | 66 | |
| NM_142045.4 | NP_650302.1 | 2 - 3 | 1441 | 66 | ||
| NM_169541.3 | NP_731840.1 | 2 - 3 | 1447 | 66 | ||
| NM_169542.3 | NP_731841.1 | 2 - 3 | 1435 | 66 | ||
| NM_169543.3 | NP_731842.1 | 2 - 3 | 1441 | 66 | ||
| NM_169544.2 | NP_731843.1 | 2 - 3 | 1587 | 66 | ||
| NM_169545.3 | NP_731844.1 | 2 - 3 | 1441 | 66 | ||
| NM_169546.3 | NP_731845.1 | 2 - 3 | 1435 | 66 | ||
| NM_169547.3 | NP_731846.1 | 2 - 3 | 1447 | 66 | ||
| GenBank mRNA | AY069241.1 | - | 2 - 3 | 1393 | 66 | |
More Information
Set Membership: |
|

Lkb1
Genomic Map