Assay Details
| Gene Symbol: omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275613.1 | NP_001262542.1 | 3 - 4 | 1283 | 104 | |
| NM_001275614.1 | NP_001262543.1 | 3 - 4 | 1283 | 104 | ||
| NM_142046.2 | NP_650303.1 | 4 - 5 | 1447 | 104 | ||
| GenBank mRNA | BT011415.1 | - | 4 - 5 | 1447 | 104 | |
More Information
Set Membership: |
|

omd
Genomic Map