Assay Details
| Gene Symbol: Adgf-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142051.3 | NP_650308.3 | 1 - 2 | 240 | 61 | |
| GenBank mRNA | AF337552.1 | - | 1 - 2 | 1124 | 61 | |
More Information
Set Membership: |
|

Adgf-C
Genomic Map