Assay Details
| Gene Symbol: kmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104307.3 | NP_001097777.1 | 7 - 8 | 4534 | 59 | |
| NM_001104308.3 | NP_001097778.1 | 7 - 8 | 4534 | 59 | ||
| NM_001144575.2 | NP_001138047.1 | 6 - 7 | 4462 | 59 | ||
| NM_001275619.2 | NP_001262548.1 | 6 - 7 | 4135 | 59 | ||
| NM_001275622.1 | NP_001262551.1 | 7 - 8 | 4534 | 59 | ||
| NM_001275623.1 | NP_001262552.1 | 6 - 7 | 4197 | 59 | ||
| NM_142053.2 | NP_650310.2 | 8 - 9 | 5367 | 59 | ||
More Information
Set Membership: |
|

kmr
Genomic Map