Assay Details
| Gene Symbol: kmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104305.3 | NP_001097775.2 | 12 - 13 | 9868 | 91 | |
| NM_001104306.3 | NP_001097776.1 | 11 - 12 | 6520 | 91 | ||
| NM_001104307.3 | NP_001097777.1 | 12 - 13 | 6658 | 91 | ||
| NM_001104308.3 | NP_001097778.1 | 12 - 13 | 6658 | 91 | ||
| NM_001144575.2 | NP_001138047.1 | 11 - 12 | 6586 | 91 | ||
| NM_001275619.2 | NP_001262548.1 | 11 - 12 | 6259 | 91 | ||
| NM_001275620.1 | NP_001262549.1 | 11 - 12 | 6520 | 91 | ||
| NM_001275622.1 | NP_001262551.1 | 12 - 13 | 6658 | 91 | ||
| NM_001275623.1 | NP_001262552.1 | 11 - 12 | 6321 | 91 | ||
| NM_142053.2 | NP_650310.2 | 13 - 14 | 7491 | 91 | ||
More Information
Set Membership: |
|

kmr
Genomic Map