Assay Details
| Gene Symbol: rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142068.2 | NP_650325.1 | 6 - 7 | 1106 | 61 | |
| NM_142069.4 | NP_650326.3 | 5 - 6 | 2463 | 61 | ||
| NM_169564.2 | NP_731875.2 | 5 - 6 | 1266 | 61 | ||
| NM_169565.2 | NP_731876.2 | 5 - 6 | 1266 | 61 | ||
| NM_169566.2 | NP_731877.1 | 6 - 7 | 1285 | 61 | ||
| GenBank mRNA | AY121682.1 | - | 6 - 7 | 1283 | 61 | |
| BT012443.1 | - | 6 - 7 | 1096 | 61 | ||
| BT021321.1 | - | 5 - 6 | 2462 | 61 | ||
| BT099706.1 | - | 5 - 6 | 1264 | 61 | ||
More Information
Set Membership: |
|

rdx
Genomic Map