Assay Details
| Gene Symbol: MetRS-m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275635.2 | NP_001262564.1 | 3 - 4 | 880 | 124 | |
| NM_001316578.1 | NP_001303507.1 | 3 - 4 | 880 | 124 | ||
| NM_142091.5 | NP_650348.1 | 3 - 4 | 880 | 124 | ||
| GenBank mRNA | AY069128.1 | - | 3 - 4 | 851 | 124 | |
More Information
Set Membership: |
|

MetRS-m
Genomic Map