Assay Details
| Gene Symbol: IFT54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142096.3 | NP_650353.1 | 1 - 2 | 165 | 80 | |
| GenBank mRNA | BT046148.1 | - | 2 - 3 | 1704 | 80 | |
More Information
Set Membership: |
|

IFT54
Genomic Map