Assay Details
| Gene Symbol: HtrA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275636.1 | NP_001262565.1 | 1 - 2 | 897 | 60 | |
| NM_142109.2 | NP_650366.1 | 1 - 2 | 645 | 60 | ||
| GenBank mRNA | AB112473.1 | - | 1 - 2 | 591 | 60 | |
| AY075206.1 | - | 1 - 2 | 645 | 60 | ||
More Information
Set Membership: |
|

HtrA2
Genomic Map