Assay Details
| Gene Symbol: Pde6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275637.2 | NP_001262566.1 | 5 - 6 | 2143 | 61 | |
| NM_001300398.1 | NP_001287327.1 | 5 - 6 | 1784 | 61 | ||
| NM_142112.5 | NP_650369.3 | 5 - 6 | 2397 | 61 | ||
More Information
Set Membership: |
|

Pde6
Genomic Map