Assay Details
| Gene Symbol: Nup93-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142147.4 | NP_650404.1 | 1 - 2 | 248 | 90 | |
| GenBank mRNA | AY058244.1 | - | 1 - 2 | 206 | 90 | |
More Information
Set Membership: |
|

Nup93-2
Genomic Map