Assay Details
| Gene Symbol: Sdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300402.1 | NP_001287331.1 | 2 - 3 | 1328 | 80 | |
| NM_142151.2 | NP_650408.1 | 2 - 3 | 1328 | 80 | ||
| GenBank mRNA | BT133291.1 | - | 2 - 3 | 1277 | 80 | |
More Information
Set Membership: |
|

Sdr
Genomic Map