Assay Details
| Gene Symbol: GlyS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275654.1 | NP_001262583.1 | 4 - 5 | 2027 | 61 | |
| NM_142165.3 | NP_650422.1 | 4 - 5 | 2683 | 61 | ||
| NM_169610.2 | NP_731967.2 | 4 - 5 | 2138 | 61 | ||
| NM_169611.2 | NP_731968.1 | 4 - 5 | 2041 | 61 | ||
| GenBank mRNA | AY052005.1 | - | 4 - 5 | 2683 | 61 | |
| BT124863.1 | - | 4 - 5 | 2117 | 61 | ||
More Information
Set Membership: |
|

GlyS
Genomic Map