Assay Details
| Gene Symbol: Gyc88E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043246.4 | NP_001036711.1 | 13 - 14 | 3050 | 69 | |
| NM_001260186.2 | NP_001247115.1 | 12 - 13 | 2956 | 69 | ||
More Information
Set Membership: |
|

Gyc88E
Genomic Map