Assay Details
| Gene Symbol: Gyc88E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043246.4 | NP_001036711.1 | 5 - 6 | 901 | 84 | |
| NM_001260186.2 | NP_001247115.1 | 4 - 5 | 807 | 84 | ||
| NM_169616.4 | NP_731974.4 | 4 - 5 | 807 | 84 | ||
More Information
Set Membership: |
|

Gyc88E
Genomic Map