Assay Details
| Gene Symbol: tefu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043247.2 | NP_001036712.1 | 18 - 19 | 4583 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tefu| Gene Symbol: tefu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043247.2 | NP_001036712.1 | 18 - 19 | 4583 | 113 | |
Set Membership: |
|