Assay Details
| Gene Symbol: CG5044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142196.3 | NP_650453.3 | 2 - 3 | 972 | 59 | |
| NM_169649.2 | NP_732020.2 | 2 - 3 | 975 | 59 | ||
| GenBank mRNA | AY052009.1 | - | 2 - 3 | 924 | 59 | |
| BT126275.1 | - | 2 - 3 | 916 | 59 | ||
More Information
Set Membership: |
|

CG5044
Genomic Map