Assay Details
| Gene Symbol: CG34404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142199.4 | NP_650456.2 | 2 - 3 | 768 | 72 | |
| NM_142200.4 | NP_650457.2 | 3 - 4 | 1103 | 72 | ||
| NM_169651.4 | NP_732023.2 | 3 - 4 | 743 | 72 | ||
| GenBank mRNA | BT133097.1 | - | 3 - 4 | 1085 | 72 | |
More Information
Set Membership: |
|

CG34404
Genomic Map