Assay Details
| Gene Symbol: Uros2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142232.2 | NP_650489.1 | 1 | 825 | 149 | |
| GenBank mRNA | BT070228.1 | - | 1 | 794 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uros2| Gene Symbol: Uros2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142232.2 | NP_650489.1 | 1 | 825 | 149 | |
| GenBank mRNA | BT070228.1 | - | 1 | 794 | 149 | |
Set Membership: |
|