Assay Details
| Gene Symbol: c(3)G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142233.3 | NP_650490.1 | 5 - 6 | 1375 | 135 | |
| NM_169670.3 | NP_732065.1 | 5 - 6 | 1375 | 135 | ||
| NM_169671.3 | NP_732066.1 | 5 - 6 | 1387 | 135 | ||
| GenBank mRNA | BT044078.1 | - | 5 - 6 | 1379 | 135 | |
More Information
Set Membership: |
|

c(3)G
Genomic Map