Assay Details
| Gene Symbol: alpha-Man-IIb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275685.1 | NP_001262614.1 | 4 - 5 | 3536 | 64 | |
| NM_001300412.1 | NP_001287341.1 | 4 - 5 | 3261 | 64 | ||
| NM_142237.3 | NP_650494.2 | 4 - 5 | 3434 | 64 | ||
| GenBank mRNA | BT150367.1 | - | 4 - 5 | 3420 | 64 | |
More Information
Set Membership: |
|

alpha-Man-IIb
Genomic Map