Assay Details
| Gene Symbol: Hmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142246.3 | NP_650503.1 | 2 - 3 | 455 | 90 | |
| GenBank mRNA | AY084209.1 | - | 2 - 3 | 457 | 90 | |
| EU571211.1 | - | 1 - 2 | 298 | 90 | ||
More Information
Set Membership: |
|

Hmt-1
Genomic Map