Assay Details
| Gene Symbol: Hmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142246.3 | NP_650503.1 | 5 - 6 | 1390 | 68 | |
| GenBank mRNA | AY084209.1 | - | 5 - 6 | 1392 | 68 | |
| EU571211.1 | - | 4 - 5 | 1233 | 68 | ||
More Information
Set Membership: |
|

Hmt-1
Genomic Map