Assay Details
| Gene Symbol: pad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142277.3 | NP_650534.2 | 2 - 3 | 606 | 73 | |
| GenBank mRNA | BT022205.1 | - | 2 - 3 | 606 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pad| Gene Symbol: pad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142277.3 | NP_650534.2 | 2 - 3 | 606 | 73 | |
| GenBank mRNA | BT022205.1 | - | 2 - 3 | 606 | 73 | |
Set Membership: |
|