Assay Details
Gene Symbol: CG17565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142283.4 | NP_650540.1 | 3 - 4 | 1119 | 66 | |
GenBank mRNA | AY051869.1 | - | 3 - 4 | 1116 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
