Assay Details
| Gene Symbol: CG14891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142303.4 | NP_650560.1 | 1 | 314 | 88 | |
| GenBank mRNA | AY058395.1 | - | 1 | 313 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG14891| Gene Symbol: CG14891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142303.4 | NP_650560.1 | 1 | 314 | 88 | |
| GenBank mRNA | AY058395.1 | - | 1 | 313 | 88 | |
Set Membership: |
|