Assay Details
| Gene Symbol: Patr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300440.1 | NP_001287369.1 | 2 - 3 | 940 | 129 | |
| NM_142335.3 | NP_650592.1 | 2 - 3 | 940 | 129 | ||
More Information
Set Membership: |
|

Patr-1
Genomic Map